missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPP25 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £366.00
Specifications
| Antigen | RPP25 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602922
|
Novus Biologicals
NBP2-94328-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653662
|
Novus Biologicals
NBP2-94328-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPP25 Polyclonal antibody specifically detects RPP25 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| RPP25 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Endocrinology | |
| PBS (pH 7.3), 50% glycerol | |
| 54913 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| FLJ20374, ribonuclease P 25kDa subunit, ribonuclease P protein subunit p25, ribonuclease P/MRP 25kDa subunit, RNase P protein subunit p25 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-199 of human RPP25 (NP_060263.2). MENFRKVRSEEAPAGCGAEGGGPGSGPFADLAPGAVHMRVKEGSKIRNLMAFATASMAQPATRAIVFSGCGRATTKTVTCAEILKRRLAGLHQVTRLRYRSVREVWQSLPPGPTQGQTPGEPAASLSVLKNVPGLAILLSKDALDPRQPGYQPPNPHPGPSSPPAAPASKRSLGEPAAGEGSAKRSQPEPGVADEDQTA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title