missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPP21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113486
This item is not returnable.
View return policy
Description
RPP21 Polyclonal Antibody for Western Blot, ELISA
RPP21 is a protein subunit of nuclear ribonuclease P, which processes the 5-prime leader sequence of precursor tRNAs.
Specifications
| RPP21 | |
| Polyclonal | |
| Unconjugated | |
| Rpp21 | |
| 0610037N01Rik; C6orf135; Cat60; Flj22638; ribonuclease P 21 subunit; ribonuclease P 21kDa subunit; ribonuclease P protein subunit p21; Ribonuclease P/MRP 21 kDa subunit; ribonuclease P/MRP 21 subunit; ribonuclease P/MRP 21kDa subunit; ribonuclease P/MRP subunit p21; ribonucleoprotein V; RNaseP protein p21; Rpp21 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 79897 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9H633 | |
| Rpp21 | |
| Recombinant Human Ribonuclease P protein subunit p21 protein (2-154AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction