missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10576-1-AP-20UL
This item is not returnable.
View return policy
Description
RNP2 a type I integral membrane protein found only in the rough endoplasmic reticulum. The encoded protein is part of an N-oligosaccharyl transferase complex that links high mannose oligosaccharides to asparagine residues found in the Asn-X-Ser/Thr consensus motif of nascent polypeptide chains. This protein is similar in sequence to the yeast oligosaccharyl transferase subunit SWP1.Specifications
| RPN2 | |
| Polyclonal | |
| Unconjugated | |
| RPN2 | |
| RIBIIR, Ribophorin 2, ribophorin II, RPN II, RPN2, RPNII, SWP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20014, 6185, 64701 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P04844, P25235, Q9DBG6 | |
| RPN2 | |
| RPN2 Fusion Protein Ag0862 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu