missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribophorin I Monoclonal Antibody (OTI6A9)

Ribophorin I Monoclonal Antibody (OTI6A9)
Brand: Invitrogen MA525711
This item is not returnable.
View return policy
Description
Ribophorin I Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
| Ribophorin I | |
| Monoclonal | |
| 0.58 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| E2RQ08, P04843 | |
| RPN1 | |
| Full length human recombinant protein of RPN1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Canine | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI6A9 | |
| Unconjugated | |
| RPN1 | |
| AU018702; D6Wsu137e; Dolichyl-diphosphooligosaccharide--protein; dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1; oligosaccharyltransfe; oligosaccharyltransferase 1 homolog; oligosaccharyltransferase complex subunit (non-catalytic); OST1; QccE-21639; QtsA-18349; RBPH1; RIBI; ribophorin I; Ribophorin-1; RPN1; Rpn-1; RPN-I; unnamed protein product | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 476516, 6184 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction