missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12894-1-AP-20UL
This item is not returnable.
View return policy
Description
Substrate-binding subunit of tRNA (adenine-N(1)-)-methyltransferase, which catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in initiator methionyl-tRNA.Specifications
| RPN1 | |
| Polyclonal | |
| Unconjugated | |
| RPN1 | |
| hrd2, OST1, RBPH1, Ribophorin 1, ribophorin I, RPN I, RPN1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 103963, 25596, 6184 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04843, P07153, Q91YQ5 | |
| RPN1 | |
| RPN1 Fusion Protein Ag3576 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction