missing translation for 'onlineSavingsMsg'
Learn More

RPMI 1640 Medium, Powder, MP Biomedicals

Product Code. 11470315 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 x 1 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470315 10 x 1 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11470315 Supplier MP Biomedicals Supplier No. 091060120
  • ONLINE EXCLUSIVE PRICE
    £154.00
    £143.85 / 10 L
    Save £10.15
    6% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent.

Specifications

Form Powder
Without Additives No Sodium Bicarbonate
Quantity 10 x 1 L
Content And Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.