Learn More
MP Biomedicals™ RPMI 1640 Medium, 1X Liq. without L-Glutamine, L-Cysteine, L-Cystine and L-Methionine
Shop All MP Biomedicals ProductsDescription
RPMI 1640 Medium was originally developed by Moore and his colleagues at Roswell Park Memorial Institute (RPMI). It was based on the RPMI 1630 line of media which utillized a bicarbonate buffering system and alterations in the amounts of amino acids and vitamins.
RPMI has successfully been used for the cultivation of normal human and neoplastic leukocytes. It is now a popular general purpose medium when properly supplemented.
Specifications
Specifications
| Form | Liquid |
| Product Type | RPMI 1640 |
| Sterility | Sterile |
| Without Additives | No Glutamine, No Cystine, No Cysteine, No Methionine |
| Quantity | 500 mL |
| pH | 6.8 to 7.4 |
| Content And Storage | 4°C |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.