missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rpmH/L34 Antibody - N-Terminus - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £351.00
Specifications
| Antigen | rpmH/L34 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5 ug/ml, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622094
|
Novus Biologicals
NBP3-23515-0.1mg |
0.1 mg |
£351.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662705
|
Novus Biologicals
NBP3-23515-0.025mg |
0.025 mg |
£153.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
rpmH/L34 Polyclonal antibody specifically detects rpmH/L34 in E. coli samples. It is validated for Western Blot, ELISASpecifications
| rpmH/L34 | |
| Western Blot 0.5 ug/ml, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS | |
| 948216 | |
| IgG | |
| Affinity purified |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| E. coli | |
| 50S ribosomal subunit protein L34, ECK3695, Large ribosomal subunit protein bL34, rimA, rpmH, ssaF | |
| Antibody was raised against an 17 amino acid peptide near the amino terminus of E. coli rpmH/L34. The immunogen is located within the first 30 amino acids of rpmH/L34. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title