missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPLP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | RPLP2 |
|---|---|
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18312884
|
Novus Biologicals
NBP3-04413-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18341917
|
Novus Biologicals
NBP3-04413-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPLP2 Polyclonal specifically detects RPLP2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| RPLP2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 6181 | |
| IgG | |
| Affinity purified |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| acidic ribosomal phosphoprotein P2, MGC71408,60S acidic ribosomal protein P2, P2, Renal carcinoma antigen NY-REN-44, ribosomal protein, large, P2, RPP2D11S2243E | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-115 of human RPLP2 (NP_000995.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title