Learn More
Invitrogen™ RPLP1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA551140
Product Code. 15625065
  • £441.00 / 100µL
Description
The antibody detects endogenous levels of total RPLP1 protein.
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein LP1 (ribosomal protein, large, P1), also known as 60S acidic ribosomal protein P1, RPP1 or RPLP1, is a 114 amino acid acidic protein that belongs to the ribosomal protein L12P family. A key component during the elongation step of protein synthesis, Ribosomal Protein LP1 forms a pentameric complex (in conjunction with LP2 and LP0) that regulates ribosome assembly and plays a role in translation initiation. Like other mammalian ribosomal proteins, Ribosomal Protein LP1 exists as multiple processed pseudogenes that are found throughout the genome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RPLP1
Polyclonal
Unconjugated
RPLP1
2410042H16Rik; 60S acidic ribosomal protein P1; acidic ribosomal phosphoprotein P1; Arpp1; C430017H15Rik; L RPP1; Large ribosomal subunit protein P1; LP1; P1; ribosomal protein lateral stalk subunit P1; ribosomal protein, large, P1; RPLP1; RPP1; RRP1
Rabbit
Antigen affinity chromatography
RUO
6176
-20°C
Liquid
Immunohistochemistry, Western Blot
1.4 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P05386
RPLP1
Fusion protein of human RPLP1.