missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPLP1 Monoclonal Antibody (1G10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524411
This item is not returnable.
View return policy
Description
Peptide Sequence: MASVSELACI YSALILHDDE VTVTEDKINA LIKAAGVNVE PFWPGLFAKA LANVNIGSLI CNVGAGGPAP AAGAAPAGGP APSTAAAPAE EKKVEAKKEE SEESDDDMGF GLFD.
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein LP1 (ribosomal protein, large, P1), also known as 60S acidic ribosomal protein P1, RPP1 or RPLP1, is a 114 amino acid acidic protein that belongs to the ribosomal protein L12P family. A key component during the elongation step of protein synthesis, Ribosomal Protein LP1 forms a pentameric complex (in conjunction with LP2 and LP0) that regulates ribosome assembly and plays a role in translation initiation. Like other mammalian ribosomal proteins, Ribosomal Protein LP1 exists as multiple processed pseudogenes that are found throughout the genome.
Specifications
| RPLP1 | |
| Monoclonal | |
| 0.47 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P05386 | |
| RPLP1 | |
| RPLP1 (NP_000994.1, 1 a.a. ~ 114 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 1G10 | |
| Unconjugated | |
| RPLP1 | |
| 2410042H16Rik; 60S acidic ribosomal protein P1; acidic ribosomal phosphoprotein P1; Arpp1; C430017H15Rik; L RPP1; Large ribosomal subunit protein P1; LP1; P1; ribosomal protein lateral stalk subunit P1; ribosomal protein, large, P1; RPLP1; RPP1; RRP1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 6176 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction