missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPLP0 Recombinant Rabbit Monoclonal Antibody (K01_3K33)

Product Code. 30825559
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825559 100 μL
30828134 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825559 Supplier Invitrogen™ Supplier No. MA564347

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

RPLP0 Recombinant Monoclonal Antibody for Western Blot

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein, which is the functional equivalent of the E. coli L10 ribosomal protein, belongs to the L10P family of ribosomal proteins. It is a neutral phosphoprotein with a C-terminal end that is nearly identical to the C-terminal ends of the acidic ribosomal phosphoproteins P1 and P2. The P0 protein can interact with P1 and P2 to form a pentameric complex consisting of P1 and P2 dimers, and a P0 monomer. The protein is located in the cytoplasm. Transcript variants derived from alternative splicing exist; they encode the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPLP0
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_3K33
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene RPLP0
Gene Accession No. P05388, P14869, P19945
Gene Alias 36B4; 60S acidic ribosomal protein P0; 60S acidic ribosomal protein P0-like protein; 60S ribosomal protein L10E; acidic ribosomal phophoprotein; acidic ribosomal phophoprotein P0; acidic ribosomal phosphoprotein P0; acidic ribosomal phosphoprotein PO; arbp; arp; fa56b12; fb04a12; L10E; Large ribosomal subunit protein uL10; LP0; MGC107165; MGC107166; MGC111226; MGC88175; neutral ribosomal phosphoprotein P0; OTTMUSP00000015585; P0; PRLP0; ribosomal protein lateral stalk subunit P0; ribosomal protein P0; ribosomal protein, large, P0; RPLP0; rplp0 protein; rplRP; RPP0; similar to acidic ribosomal phosphoprotein PO; wu:fa56b12; wu:fb15a08; wu:fk48c12
Gene Symbols RPLP0
Host Species Rabbit
Immunogen Recombinant protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11837, 6175, 64205
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.