missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL7L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565623
This item is not returnable.
View return policy
Description
Immunogen sequence: HEIAFPGKHFQ EISWFLCPFH LSVARHATKN RVGFLKEMGT PGYRGERINQ LIRQLN Highest antigen sequence identity to the following orthologs - mouse 88%, rat 93%.
RPL7L1 gene ontology annotations related to this gene include blastocyst formation.
Specifications
| RPL7L1 | |
| Polyclonal | |
| Unconjugated | |
| RPL7L1 | |
| 1500016H10Rik; 60S ribosomal protein L7-like 1; dJ475N16.4; Large ribosomal subunit protein uL30-like 1; RGD1308469; ribosomal protein L7 like 1; ribosomal protein L7-like 1; RPL7L1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 285855 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6DKI1 | |
| RPL7L1 | |
| Recombinant Human RPL7L1. Recombinant protein control fragment (Product #RP-106285). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction