Learn More
RPL7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB100-2269
This item is not returnable.
View return policy
Description
RPL7 Polyclonal specifically detects RPL7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| Polyclonal | |
| humL7-1,60S ribosomal protein L7, MGC117326, ribosomal protein L7 | |
| 29 kDa | |
| 0.1 mL | |
| Human, Mouse | |
| IgG |
| Western Blot 1:2000 - 1:10000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:100-1:500, Knockdown Validated | |
| RPL7 | |
| Affinity Purified | |
| RUO | |
| Store at 4C. Do not freeze. |