missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL7 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2268H
This item is not returnable.
View return policy
Description
RPL7 Polyclonal specifically detects RPL7 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| RPL7 | |
| Polyclonal | |
| Western Blot | |
| humL7-1,60S ribosomal protein L7, MGC117326, ribosomal protein L7 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| HRP | |
| PBS with No Preservative | |
| RPL7 | |
| The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human Ribosomal Protein 7 using the numbering given in entry NP_000962.2 (GeneID 6129). | |
| 100 μL | |
| Immune System Diseases, Immunology | |
| 6129 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction