missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£361.00
Specifications
| Antigen | RPL6 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
RPL6 Polyclonal specifically detects RPL6 in Human samples. It is validated for Western Blot.Specifications
| RPL6 | |
| Polyclonal | |
| Purified | |
| RUO | |
| DNA-binding protein TAXREB107, Neoplasm-related protein C140, ribosomal protein L6,60S ribosomal protein L6, SHUJUN-2, TaxREB107, Tax-responsive enhancer element-binding protein 107, TXREB1 | |
| RPL6 | |
| IgG | |
| Protein A purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Q02878 | |
| 6128 | |
| Synthetic peptides corresponding to RPL6 (ribosomal protein L6) The peptide sequence was selected from the N terminal of RPL6. Peptide sequence AGEKVEKPDTKEKKPEAKKVDAGGKVKKGNLKAKKPKKGKPHCSRNPVLV. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title