missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL39L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549960
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total RPL39L protein.
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron containing gene within their coding regions. Ribosomal Protein L39 (RPL39), also known as 60S Ribosomal Protein L39, consists of 51 amino acids and is the smallest protein found in liver ribosomes. Ribosomal Protein L39 localizes to cytoplasm and is a member of the S39E family of ribosomal proteins. The gene encoding Ribosomal Protein L39 maps to human chromosome Xq24, and shares 92 percent homology with Ribosomal Protein L39L (Ribosomal Protein L39-like), which is also known as RPL39L, L39-2 or RPL39L1. Ribosomal Protein L39L is associated with the 60S ribosomal subunit and is expressed in testis with localization in nucleolus.
Specifications
| RPL39L | |
| Polyclonal | |
| Unconjugated | |
| RPL39L | |
| 60S ribosomal protein L39-2; 60S ribosomal protein L39-like; L39-2; Large ribosomal subunit protein eL39-like; ribosomal protein L39 like; ribosomal protein L39-like 1; ribosomal protein L39-like protein; RPL39L; RPL39L1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116832 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96EH5 | |
| RPL39L | |
| Synthesized peptide derived from internal of human RL39L (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction