missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL37A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107252
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Goat: 79%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L37AE family of ribosomal proteins. It is located in the cytoplasm. The protein contains a C4-type zinc finger-like domain. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPL37A | |
| Polyclonal | |
| Unconjugated | |
| RPL37A | |
| 60S ribosomal protein L37a; L37A; Large ribosomal subunit protein eL43; ribosomal protein L37a; ribosomal protein L37a-like; Rpl37a | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6168 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| P61513 | |
| RPL37A | |
| Synthetic peptide directed towards the middle region of human RPL37A (aa 42-92). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction