missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL37 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95175-0.1ml
This item is not returnable.
View return policy
Description
RPL37 Polyclonal antibody specifically detects RPL37 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RPL37 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp686G1699, G1.16, MGC99572,60S ribosomal protein L37,60S ribosomal protein L37a, ribosomal protein L37 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-97 of human RPL37 (NP_000988.1). MTKGTSSFGKRRNKTHTLCRRCGSKAYHLQKSTCGKCGYPAKRKRKYNWSAKAKRRNTTGTGRMRHLKIVYRRFRHGFREGTTPKPKRAAVAASSSS | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6167 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction