missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552245
This item is not returnable.
View return policy
Description
Immunogen sequence: AKIKARDLRG KKKEELLKQL DDLKVELSQL RVAKVTGGAA SKLSKIRVVR KSIARVLTVI NQTQKENLRK FYKGKKYKPL DLRPKKTRAM RRRLNKHEEN LKTKKQQRKE RLYPLRKYAV K Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L29P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPL35 | |
| Polyclonal | |
| Unconjugated | |
| RPL35 | |
| 2410039E09Rik; 60S ribosomal protein L35; L35; Large ribosomal subunit protein uL29; ribosomal protein L35; Rpl35 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11224 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P42766 | |
| RPL35 | |
| Recombinant protein corresponding to Human RPL35. Recombinant protein control fragment (Product #RP-102211). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction