missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL34, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00006164-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: NKTRLSRTPGNRIVYLYTKKVGKAPKSACGVCPGRLRGVRAVRPKVLMRLSKTKKHVSRAYGGSMCAKCVRDRIKRAFLIEEQKIVVKVLKAQAQSQKAKSpecifications
| RPL34 | |
| Polyclonal | |
| 50 % glycerol | |
| NM_000995 | |
| RPL34 | |
| RPL34 (NP_000986, 18 a.a. ∼ 117 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 6164 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ribosomal protein L34 | |
| MGC111005 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction