missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL32 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | RPL32 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RPL32 Polyclonal specifically detects RPL32 in Mouse samples. It is validated for Western Blot.Specifications
| RPL32 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| M, ribosomal protein L32,60S ribosomal protein L32 | |
| The immunogen is a synthetic peptide directed towards the middle region of RPL32 (NP_742083). Peptide sequence GSNKKTKHMLPSGFRKFLVHNVKELEVLLMCNKSYCAEIAHNVSSKNRKA | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 6161 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title