Learn More
Invitrogen™ RPL31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585599
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (91%), Xenopus laevis (93%), Dog (100%), Chicken (98%), Rhesus Monkey (100%), Bovine (100%). Positive Control: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPL31 is a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L31E family of ribosomal proteins. It is located in the cytoplasm.
Specifications
| RPL31 | |
| Polyclonal | |
| Unconjugated | |
| RPL31 | |
| 60S ribosomal protein L31; L31; Large ribosomal subunit protein eL31; ribosomal protein L31; RPL31 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6160 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.9 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P62899 | |
| RPL31 | |
| Recombinant protein encompassing a sequence within the center region of human RPL31. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.