Learn More
Invitrogen™ RPL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA593323
Description
Immunogen sequence: MSHRKFSAPR HGSLGFLPRK RSSRHRGKVK SFPKDDPSKP VHLTAFLGYK AGMTHIVREV DRPGSKVNKK EVVEAVTIVE TPPMVVVGIV GYVETPRGLR TFKTVFAEHI SDECKRRFYK NWHKSKKKAF TKYCKKWQDE DGKKQLEKDF SSMKKYCQVI RVIAHTQMRL LPLRQKKAHL MEIQVNGGTV AEKLDWARER LEQQVPVNQV FGQDEMIDVI GVTKGKGYKG VTSRWHTKKL PRKTHRGLRK VACIGAWHPA; Positive Samples: U-87MG; Cellular Location: Cytoplasm, Nucleus, nucleolus.
This gene encodes a member of the SWI2/SNF2 family of proteins, which play a critical role in altering protein-DNA interactions. The encoded protein has been shown to have dsDNA-dependent ATPase activity and RNA polymerase II termination activity. This protein interacts with cell division cycle 5-like, associates with human splicing complexes, and plays a role in pre-mRNA splicing.
Specifications
| RPL3 | |
| Polyclonal | |
| Unconjugated | |
| RPL3 | |
| 60S ribosomal protein L3; ASC-1; F2; HIV-1 TAR RNA-binding protein B; J1; J1 protein; L3; L4; Large ribosomal subunit protein uL3; OK/SW-cl.32; ribosomal protein L3; ribosomal protein L3 homolog; ribosomal protein L3, isoform a; RL3; RPL3; TARBP-B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27367, 6122 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.62 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P27659, P39023 | |
| RPL3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human RPL3 (NP_0009581). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.