missing translation for 'onlineSavingsMsg'
Learn More

RPL3 Mouse anti-Human, Mouse, Rat, Clone: 2G1B8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66130-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877725

  • £130.00 / 20µL
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ribosomes, the complexes that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L3P family of ribosomal proteins and it is located in the cytoplasm. The protein can bind to the HIV-1 TAR mRNA, and it has been suggested that the protein contributes to tat-mediated transactivation. This gene is co-transcribed with several small nucleolar RNA genes, which are located in several of this gene′s introns. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications

Specifications

RPL3
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P21531, P27659, P39023
RPL3
RPL3 Fusion Protein Ag20400
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
2G1B8
Unconjugated
RPL3
60S ribosomal protein L3, ribosomal protein L3, RPL3, TARBP B
Mouse
Protein G
RUO
27367, 300079, 6122
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers