Learn More
RPL28 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16649-1-AP-20UL
 View more versions of this product
Product Code. 16836344
  • £135.00 / 20µL
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L28E family of ribosomal proteins. It is located in the cytoplasm. Variable expression of this gene in colorectal cancers compared to adjacent normal tissues has been observed, although no correlation between the level of expression and the severity of the disease has been found. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
Specifications
RPL28
Polyclonal
Unconjugated
RPL28
60S ribosomal protein L28, ribosomal protein L28, RPL28
Rabbit
Antigen Affinity Chromatography
RUO
19943, 6158, 64638
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunofluorescence
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P17702, P41105, P46779
RPL28
RPL28 Fusion Protein Ag10036
20 μL
Primary
Rat, Mouse, Human