Learn More
RPL27A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16002-1-AP-20UL
 View more versions of this product
Product Code. 16895434
  • £135.00 / 20µL
Description
Rpl27a gene ontology annotations related to this gene include translation.
Specifications
Specifications
RPL27A
Polyclonal
Unconjugated
RPL27A
60S ribosomal protein L27a, ribosomal protein L27a, RPL27A
Rabbit
Antigen Affinity Chromatography
RUO
26451, 293418, 6157
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P14115, P18445, P46776
RPL27A
RPL27A Fusion Protein Ag8596
20 μL
Primary
Human, Rat, Mouse