missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL27 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570189
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 85%; Zebrafish: 93%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L27E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L27E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPL27 | |
| Polyclonal | |
| Unconjugated | |
| RPL27 | |
| 60S ribosomal protein L27; B173; fc93a08; L27; Large ribosomal subunit protein eL27; ribosomal protein L27; rpl27; similar to ribosomal protein L27; wu:fc93a08; zgc:56171 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19942 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P61358 | |
| RPL27 | |
| synthetic peptide directed towards the middle region of human RPL27 (aa 30-79). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction