missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL22L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584777
This item is not returnable.
View return policy
Description
Immunogen sequence: QFSKRYLKYL TKKYLKKNNL RDWLRVVAS Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
Ribosomal Protein L22 is also known as heparin-binding protein HBp15, because it binds heparin in the submandibular gland and brain. This small protein is also associated with two small nuclear RNAs called EBERs (Epstein- Barr encoded RNAs). These RNAs are synthesized in large amounts by human B lymphocytes infected with Epstein-Barr virus (EBV). Ribosomal protein L22, like L4, contains a globular domain that sits on the surface of the large ribosomal subunit and an extended loop that penetrates its core. These extensions contact multiple domains of 23S rRNA, indicating a potential, but not essential, role in rRNA folding during ribosomal assembly.
Specifications
| RPL22L1 | |
| Polyclonal | |
| Unconjugated | |
| RPL22L1 | |
| 60S ribosomal protein L22-like 1; EAP; Large ribosomal subunit protein eL22-like 1; RGD1309517; ribosomal protein L22 like 1; ribosomal protein L22-like 1; RPL22; RPL22L1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 200916 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P5R6 | |
| RPL22L1 | |
| Recombinant protein corresponding to Human RPL22L1. Recombinant protein control fragment (Product #RP-108539). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction