missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15226-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the protein kinase C and casein kinase substrate in neurons family. The encoded protein is involved in linking the actin cytoskeleton with vesicle formation. Alternative splicing results in multiple transcript variants.Specifications
| RPL21 | |
| Polyclonal | |
| Unconjugated | |
| RPL21 | |
| 60S ribosomal protein L21, L21, ribosomal protein L21, RPL21 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19933, 6144, 79449 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O09167, P20280, P46778 | |
| RPL21 | |
| RPL21 Fusion Protein Ag7444 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction