missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112700
This item is not returnable.
View return policy
Description
RPL19 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L19E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPL19 | |
| Polyclonal | |
| Unconjugated | |
| RPL19 | |
| 60S ribosomal protein L19; DKFZp779D216; FLJ27452; HGNC:10312; L19; Large ribosomal subunit protein eL19; MGC71997; ribosomal protein L19; RPL19 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein G | |
| RUO | |
| 19921, 6143 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P84098, P84099 | |
| RPL19 | |
| Recombinant Human 60S ribosomal protein L19 protein (79-196AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction