missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL18 Polyclonal Antibody

RPL18 Polyclonal Antibody
Brand: Invitrogen PA588025
This item is not returnable.
View return policy
Description
RPL18 Polyclonal Antibody for Western Blot
Specifications
| RPL18 | |
| Polyclonal | |
| Unconjugated | |
| RPL18 | |
| 60S ribosomal protein L18; L18; Large ribosomal subunit protein eL18; ribosomal protein L18; RPL18; Rpl18a | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19899, 6141 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.21 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P35980, Q07020 | |
| RPL18 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-188 of human RPL18 (NP_0009701). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction