Learn More
Invitrogen™ RPL13 Recombinant Rabbit Monoclonal Antibody (24GB770)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557226
 View more versions of this product
Product Code. 30275100
  • £126.00 / 20µL
Description
RPL13 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPL13 is a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L13E family of ribosomal proteins. It is located in the cytoplasm. This gene is expressed at significantly higher levels in benign breast lesions than in breast carcinomas. Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L13E family of ribosomal proteins. It is located in the cytoplasm. This gene is expressed at significantly higher levels in benign breast lesions than in breast carcinomas. Transcript variants derived from alternative splicing and/or alternative polyadenylation exist; these variants encode the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RPL13
Recombinant Monoclonal
Unconjugated
RPL13
60S ribosomal protein L13; A52; BBC1; breast basic conserved protein 1; D16S444E; D16S44E; FLJ27453; FLJ27454; L13; Large ribosomal subunit protein eL13; LOC508221 protein; MGC117342; MGC71373; OK/SW-cl.46; ribosomal protein L13; RPL13; RPL13 protein-like; zgc:92272
Rabbit
Affinity chromatography
RUO
270106, 6137, 81765
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB770
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P26373, P41123, P47963
RPL13
A synthesized peptide derived from human RPL13 (aa 1-20).
20 μL
Primary
Human, Mouse, Rat