missing translation for 'onlineSavingsMsg'
Learn More

RPL11 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18409547 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409547 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409547 Supplier Novus Biologicals Supplier No. NBP222283AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RPL11 Polyclonal antibody specifically detects RPL11 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL11
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias cell growth-inhibiting protein 34,60S ribosomal protein L11, CLL-associated antigen KW-12, DBA7, GIG34, ribosomal protein L11
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 128 and 178 of human Ribosomal Protein L11 using the numbering given in entry NP_000966.2 (GeneID 6135).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6135
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.