missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL10A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562845
This item is not returnable.
View return policy
Description
Immunogen sequence: PGLNKAGKFP SLLTHNENMV AKVDEVK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L1P family of ribosomal proteins. It is located in the cytoplasm. The expression of this gene is downregulated in the thymus by cyclosporin-A (CsA), an immunosuppressive drug. Studies in mice have shown that the expression of the ribosomal protein L10a gene is downregulated in neural precursor cells during development. This gene previously was referred to as NEDD6 (neural precursor cell expressed, developmentally downregulated 6), but it has been renamed RPL10A (ribosomal protein 10a). As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPL10A | |
| Polyclonal | |
| Unconjugated | |
| RPL10A | |
| 60S ribosomal protein L10a; CSA19; Csa-19; L10A; Large ribosomal subunit protein uL1; Nedd6; NEDD-6; Neural precursor cell expressed developmentally down-regulated protein 6; ribosomal protein L10a; RPL10A; Unknown (protein for MGC:73082); wu:fb94f08; zgc:73082; zgc:86881 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4736 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P62906 | |
| RPL10A | |
| Recombinant protein corresponding to Human RPL10A. Recombinant protein control fragment (Product #RP-99544). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction