missing translation for 'onlineSavingsMsg'
Learn More

RPL10A Antibody (OTI2G9) [PerCP], Novus Biologicals™

Product Code. 30687414 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687414 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687414 Supplier Novus Biologicals Supplier No. NBP344967PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RPL10A Monoclonal antibody specifically detects RPL10A in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL10A
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI2G9
Conjugate PerCP
Formulation PBS
Gene Alias CSA-19, Csa-19,60S ribosomal protein L10a, NEDD6, NEDD-6, Neural precursor cell expressed developmentally down-regulated protein 6, ribosomal protein L10a
Host Species Mouse
Immunogen Full length human recombinant protein of human RPL10A (NP_009035) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4736
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.