missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL10 Polyclonal Antibody

RPL10 Polyclonal Antibody
Brand: Thermo Scientific PA5104473
This item is not returnable.
View return policy
Description
RPL10 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| RPL10 | |
| Polyclonal | |
| Unconjugated | |
| Rpl10 | |
| 24.6 kDa protein; 24.6kDa protein; 60S ribosomal protein L10; AUTSX5; D0HXS648; DXHXS648; DXHXS648E; DXS648; DXS648E; L10; Laminin receptor homolog; Large ribosomal subunit protein uL16; NOV; Protein QM; Protein QM homolog; QM; QM16; ribosomal protein 10; ribosomal protein L10; Rpl10; RPLPO; Tumor suppressor QM; Wilms tumor-related protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 110954, 6134, 81764 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P27635, Q6PDV7, Q6ZWV3 | |
| Rpl10 | |
| A synthesized peptide derived from human RPL10(Accession P27635), corresponding to amino acid residues N14-A64. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction