missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL10 Monoclonal Antibody (OTI6B11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526901
This item is not returnable.
View return policy
Description
RPL10 Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a ribosomal protein that is a component of the 60S ribosome subunit. The related protein in chicken can bind to c-Jun and can repress c-Jun-mediated transcriptional activation. Some studies have detected an association between variation in this gene and autism spectrum disorders, though others do not detect this relationship. There are multiple pseudogenes of this gene dispersed throughout the genome. Alternative splicing results in multiple transcript variants.
Specifications
| RPL10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P27635 | |
| Rpl10 | |
| Full length human recombinant protein of RPL10 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6B11 | |
| Unconjugated | |
| Rpl10 | |
| 24.6 kDa protein; 24.6kDa protein; 60S ribosomal protein L10; AUTSX5; D0HXS648; DXHXS648; DXHXS648E; DXS648; DXS648E; L10; Laminin receptor homolog; Large ribosomal subunit protein uL16; NOV; Protein QM; Protein QM homolog; QM; QM16; ribosomal protein 10; ribosomal protein L10; Rpl10; RPLPO; Tumor suppressor QM; Wilms tumor-related protein | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6134 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction