missing translation for 'onlineSavingsMsg'
Learn More

RPIP8 Antibody (1A8), Novus Biologicals™

Product Code. 18325039 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325039 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325039 Supplier Novus Biologicals Supplier No. H00010900M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RPIP8 Monoclonal antibody specifically detects RPIP8 in Human, Rat samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPIP8
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1A8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH06194
Gene Alias Rap2-interacting protein 8, RAP2IPRUN domain-containing protein 3A, RPIP-8, RPIP8RaP2 interacting protein 8, RUN domain containing 3A
Host Species Mouse
Immunogen RPIP8 (AAH06194, 1 a.a. ~ 405 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEASFVQTTMALGLSSKKASSRNVAVERKNLITVCRFSVKTLLEKYTAEPIDDSSEEFVNFAAILEQILSHRFKACAPAGPVSWFSSDGQRGFWDYIRLACSKVPNNCVSSIENMENISTARAKGRAWIRVALMEKRMSEYITTALRDTRTTRRFYDSGAIMLRDEATILTGMLIGLSAIDFSFCLKGEVLDGKTPVVIDYTPYLKFTQSYDYLTDEEERYSAESSTSEDNSPEHPYLPLVTDEDSWYSKWHKMEQKFRIVYAQKGYLEELVRLRESQLKDLEAENRRLQLQLEEAAAQNQREKRELEGVILELQEQLTGLIPSDHAPLAQGSKELTTPLVNQWPSLGTLNGAEGASNSKLYRRHSFMSTEPLSAEASLSSDSQRLGEGTRDEEPWGPIGSSEPN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10900
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.