missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPH3AL Polyclonal Antibody
Description
Peptide sequence: RIYTWARGRV VSSDSDSDSD LSSSSLEDRL PSTGVRDRKG DKPWKESGGS Sequence homology: Cow: 79%; Human: 100%.
The protein encoded by this gene plays a direct regulatory role in calcium-ion-dependent exocytosis in both endocrine and exocrine cells and plays a key role in insulin secretion by pancreatic cells. This gene is likely a tumor suppressor. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
Specifications
| Antigen | RPH3AL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | RPH3AL |
| Gene Accession No. | Q9UNE2 |
| Gene Alias | Gm1753; no C2 domains protein; NOC2; rab effector Noc2; rabphilin 3A like (without C2 domains); rabphilin 3A-like (without C2 domains); rabphilin-3A-like protein; Rph3al |
| Gene Symbols | RPH3AL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?