missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPGRIP1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565271
This item is not returnable.
View return policy
Description
Immunogen sequence: TKYGNSLLEEA RGEIRNLENV IQSQRGQIEE LEHLAEILKT QLRRKENEIE LSLLQLREQQ ATDQRSNIRD NVEMIKLHKQ LVEKSNALSA MEGKFIQLQE KQRTLRIS Highest antigen sequence identity to the following orthologs - mouse 95%, rat 94%.
The protein encoded by this gene can localize to the basal body-centrosome complex or to primary cilia and centrosomes in ciliated cells. The encoded protein has been found to interact with nephrocystin-4. Defects in this gene are a cause of Joubert syndrome type 7 and Meckel syndrome type 5. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| RPGRIP1L | |
| Polyclonal | |
| Unconjugated | |
| RPGRIP1L | |
| 1700047E16Rik; 4931437C01; CORS3; DKFZp686C0668; fantom; fantom homolog; Ftm; JBTS7; KIAA1005; MKS5; Nephrocystin-8; NPHP8; PPP1R134; Protein fantom; protein phosphatase 1, regulatory subunit 134; RGD1311099; RPGR-interacting protein 1-like protein; RPGRIP1 like; RPGRIP1L; Rpgrip1-like; RPGRIP1-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23322 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.9 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q68CZ1 | |
| RPGRIP1L | |
| Recombinant Human RPGRIP1L. Recombinant protein control fragment (Product #RP-105989). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction