missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPGRIP1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | RPGRIP1L |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18227951
|
Novus Biologicals
NBP2-57686 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18636356
|
Novus Biologicals
NBP2-57686-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPGRIP1L Polyclonal specifically detects RPGRIP1L in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| RPGRIP1L | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 23322 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TKYGNSLLEEARGEIRNLENVIQSQRGQIEELEHLAEILKTQLRRKENEIELSLLQLREQQATDQRSNIRDNVEMIKLHKQLVEKSNALSAMEGKFIQLQEKQRTLRIS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| CORS3, FTMDKFZp686C0668, JBTS7fantom homolog, KIAA1005RPGRIP1-like protein, MKS5nephrocystin 8, NPHP8, protein fantom, RPGR-interacting protein 1-like protein, RPGRIP1-like | |
| RPGRIP1L | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title