missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPGR Recombinant Rabbit Monoclonal Antibody (0S8U8)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£147.00 - £363.00

Specifications

Antigen RPGR
Clone 0S8U8
Concentration 1.49 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30282858
View Documents
Invitrogen™
MA558011
20 μL
£147.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30283521
GREENER_CHOICE
View Documents
Invitrogen™
MA558012
100 μL
£363.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunogen sequence: MREPEELMPD SGAVFTFGKS KFAENNPGKF WFKNDVPVHL SCGDEHSAVV TGNNKLYMFG SNNWGQLGLG SKSAISKPTC VKALKPEKVK LAACGRNHTL VSTEGGNVYA TGGNNEGQLG LGDTEERNTF HVISFFTSEH KIKQLSAGSN TSAALTEDGR LFMWGDNSEG QIGLKNVSNV CVPQQVTIGK PVSWISCGYY HSAFVTTDGE LYVFGEPENG KLGLPNQLLG NHRTPQLVSE IPEKVIQVAC GGEHTVVLTE.

This gene encodes a protein with a series of six RCC1-like domains (RLDs), characteristic of the highly conserved guanine nucleotide exchange factors. The encoded protein is found in the Golgi body and interacts with RPGRIP1. This protein localizes to the outer segment of rod photoreceptors and is essential for their viability. Mutations in this gene have been associated with X-linked retinitis pigmentosa (XLRP). Multiple alternatively spliced transcript variants that encode different isoforms of this gene have been reported, but the full-length natures of only some have been determined.
Specifications

Specifications

RPGR
1.49 mg/mL
ELISA, Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q9R0X5
19893, 367733
Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human RPGR (NP_001030025.1).
Antibody
0S8U8
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Mouse, Rat
RPGR
COD1; CORDX1; CRD; mRpgr; orf15; PCDX; retinitis pigmentosa 15; retinitis pigmentosa 3 GTPase regulator; retinitis pigmentosa GTPase regulator; RP15; RP3; Rp3h; Rpgr; X-linked retinitis pigmentosa GTPase regulator; XLRP3
RPGR
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.