missing translation for 'onlineSavingsMsg'
Learn More

RPGR Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA567290

Product Code. 15959954

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PFFGNLPDRGM NTESEENKDF VKKRESCKQD VIFDSERESV EKPDSYMEGA SESQQGIADG FQQPEAIEFS SGEKEDDEVE TDQNIRYGRK LIEQG Highest antigen sequence identity to the following orthologs - mouse 48%, rat 29%.

This gene encodes a protein with a series of six RCC1-like domains (RLDs), characteristic of the highly conserved guanine nucleotide exchange factors. The encoded protein is found in the Golgi body and interacts with RPGRIP1. This protein localizes to the outer segment of rod photoreceptors and is essential for their viability. Mutations in this gene have been associated with X-linked retinitis pigmentosa (XLRP). Multiple alternatively spliced transcript variants that encode different isoforms of this gene have been reported, but the full-length natures of only some have been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RPGR
Polyclonal
Unconjugated
RPGR
X-linked retinitis pigmentosa GTPase regulator; RPGR; RP3; XLRP3; CRD; COD1; PCDX; RP15; orf15; CORDX1
Rabbit
Antigen Affinity Chromatography
RUO
6103
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunofluorescence
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q92834
RPGR
Recombit Human RPGR
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only