Learn More
Invitrogen™ RPGR Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA579934
Product Code. 15985495
  • £393.00 / 100µg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat liver tissue, rat kidney tissue, rat brain tissue, rat heart tissue.
This gene encodes a protein with a series of six RCC1-like domains (RLDs), characteristic of the highly conserved guanine nucleotide exchange factors. The encoded protein is found in the Golgi body and interacts with RPGRIP1. This protein localizes to the outer segment of rod photoreceptors and is essential for their viability. Mutations in this gene have been associated with X-linked retinitis pigmentosa (XLRP). Multiple alternatively spliced transcript variants that encode different isoforms of this gene have been reported, but the full-length natures of only some have been determined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RPGR
Polyclonal
Unconjugated
RPGR
COD1; CORDX1; CRD; mRpgr; orf15; PCDX; retinitis pigmentosa 15; retinitis pigmentosa 3 GTPase regulator; retinitis pigmentosa GTPase regulator; RP15; RP3; Rp3h; Rpgr; X-linked retinitis pigmentosa GTPase regulator; XLRP3
Rabbit
Antigen affinity chromatography
RUO
367733, 6103
-20°C
Lyophilized
ELISA, Western Blot
500 μg/mL
PBS with 4mg trehalose and 0.05mg sodium azide
Q92834
RPGR
E. coli-derived human RPGR recombinant protein (Position: A13-F338).
100 μg
Primary
Human, Rat