missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPE65 Antibody (401.8B11.3D9) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-355JF525
This item is not returnable.
View return policy
Description
RPE65 Monoclonal antibody specifically detects RPE65 in Bovine,Canine,Chicken,Human,Mouse,Porcine,Primate,Rat,Xenopus samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),In vivo Assay,Simple Western,Immunofluorescence,CyTOF,Gene Knock-Out,Immunocytochemistry/ Immunofluorescence
Specifications
| RPE65 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| All-trans-retinyl-palmitate hydrolase, BCO family, member 3, BCO3, EC 3.1.1.64, EC:3.1.1.64, EC:5.3.3.22, LCA2, lutein isomerase, meso-zeaxanthin isomerase, mRPE65, p63, RBP-binding membrane protein, rd12, retinal pigment epithelium specific protein 65, Retinal pigment epithelium-specific 65 kDa protein, retinal pigment epithelium-specific protein 65kDa, retinitis pigmentosa 20 (autosomal recessive), retinoid isomerohydrolase, Retinol isomerase, RP20, RPE65, RPE65, retinoid isomerohydrolase, sRPE65 | |
| This RPE65 Antibody (401.8B11.3D9) was developed against bovine RPE65 microsomal membrane proteins. [UniProt# Q28175] | |
| 0.2 mL | |
| Cellular Markers, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Sensory Systems, Signal Transduction, Vision | |
| 6121 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vivo Assay, Simple Western, Immunofluorescence, CyTOF, Gene Knock-Out, Immunocytochemistry/Immunofluorescence | |
| 401.8B11.3D9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bovine, Canine, Chicken, Human, Mouse, Pig, Primate, Rat, Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction