missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPC62 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35239-20ul
This item is not returnable.
View return policy
Description
RPC62 Polyclonal antibody specifically detects RPC62 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| RPC62 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| DNA-directed III 62 kDa polypeptide, DNA-directed RNA polymerase III subunit C, DNA-directed RNA polymerase III subunit RPC3, polymerase (RNA) III (DNA directed) polypeptide C (62kD), RNA polymerase III 62 kDa subunit, RNA polymerase III subunit C3, RPC62RPC3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human RPC62 (NP_006459.3).,, Sequence:, MTQAEIKLCSLLLQEHFGEIVEKIGVHLIRTGSQPLRVIAHDTGTSLDQVKKALCVLVQHNLVSYQVHKRGVVEYEAQCSRVLRMLRYPRYIYTTKTLYSDTGELIVEELLLNGKLTMSAVVKKVADRLTETMEDGKTMDYAEVSNTFVRLADTHFVQRCPSVPTTENSDPGPPPPAPTLVINEKDMYLVPKLSLIGKGKRRRSSDEDAAGEPKAKRPKYTTDNKEPIPDDGIYWQANLDRFHQHFRDQAIVSAVANRMDQTSSEIVRTM | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10623 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction