missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RPC62 Polyclonal antibody specifically detects RPC62 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | RPC62 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DNA-directed III 62 kDa polypeptide, DNA-directed RNA polymerase III subunit C, DNA-directed RNA polymerase III subunit RPC3, polymerase (RNA) III (DNA directed) polypeptide C (62kD), RNA polymerase III 62 kDa subunit, RNA polymerase III subunit C3, RPC62RPC3 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 484 and 534 of human Polymerase (RNA) III (DNA directed) Polypeptide C (62kD) using the numbering given in entry NP_006459.3 (GeneID 10623). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?