missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPB9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | RPB9 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18283902
|
Novus Biologicals
NBP2-58387 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647366
|
Novus Biologicals
NBP2-58387-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPB9 Polyclonal specifically detects RPB9 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| RPB9 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 5438 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DELTQIIADVSQDPTLPRTEDHPCQKCGHKEAVFFQSHSARAEDAMRLYYVCT | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| DNA-directed RNA polymerase II 14.5 kDa polypeptide, DNA-directed RNA polymerase II subunit I, DNA-directed RNA polymerase II subunit RPB9, hRPB14.5, polymerase (RNA) II (DNA directed) polypeptide I (14.5kD), polymerase (RNA) II (DNA directed) polypeptide I, 14.5kDa, RNA polymerase II 14.5 kDa subunit, RNA polymerase II subunit B9, RPB14.5, RPB9 | |
| POLR2I | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title