missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPB9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£222.00
Specifications
| Antigen | RPB9 |
|---|---|
| Dilution | Western Blot 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RPB9 Polyclonal specifically detects RPB9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RPB9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5438 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PDGTYEPGFVGIRFCQECNNMLYPKEDKENRILLYACRNCDYQQEADNSCIYVNKITHEVDELTQIIADVSQDPTLPRTEDHPCQKCGHKEAVFFQSHSA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| DNA-directed RNA polymerase II 14.5 kDa polypeptide, DNA-directed RNA polymerase II subunit I, DNA-directed RNA polymerase II subunit RPB9, hRPB14.5, polymerase (RNA) II (DNA directed) polypeptide I (14.5kD), polymerase (RNA) II (DNA directed) polypeptide I, 14.5kDa, RNA polymerase II 14.5 kDa subunit, RNA polymerase II subunit B9, RPB14.5, RPB9 | |
| POLR2I | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title